missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylacetamide 98.0+%, TCI America™

$231.14 - $1181.81

Chemical Identifiers

CAS 588-46-5
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.193
MDL Number MFCD00059204
InChI Key UZJLYRRDVFWSGA-UHFFFAOYSA-N
Synonym N-Acetylbenzylamine
PubChem CID 11500
IUPAC Name N-benzylacetamide
SMILES CC(=O)NCC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B040425G
View Documents
TCI America
B040425G
25 g
Each for $231.14
Only null left
 
B0404500G
View Documents
TCI America
B0404500G
500 g
Each for $1,181.81
Only null left
 
Description

Description

Specifications

Chemical Identifiers

588-46-5
149.193
UZJLYRRDVFWSGA-UHFFFAOYSA-N
11500
CC(=O)NCC1=CC=CC=C1
C9H11NO
MFCD00059204
N-Acetylbenzylamine
N-benzylacetamide

Specifications

588-46-5
White-Yellow
C9H11NO
25 g
UZJLYRRDVFWSGA-UHFFFAOYSA-N
N-benzylacetamide
11500
≥98.0% (GC)
N-Benzylacetamide
61°C
157°C
MFCD00059204
N-Acetylbenzylamine
CC(=O)NCC1=CC=CC=C1
149.193
149.19
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0373&(9)-0024

RTECSNumber : AB4546300

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'