missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-N-(méthoxyméthylle)-N-triméthylsilylméthylamine 98,0+%, TCI America™

Catalog No. B19385G
Change view
Click to view available options
Quantité:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
B19385G 5 g
B193825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19385G Supplier TCI America Supplier No. B19385G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 93102-05-7
Formule moléculaire C13H23NOSi
Poids moléculaire (g/mol) 237.418
Numéro MDL MFCD00674005
Clé InChI RPZAAFUKDPKTKP-UHFFFAOYSA-N
Synonyme n-methoxymethyl-n-trimethylsilylmethyl benzylamine, n-benzyl-1-methoxy-n-trimethylsilyl methyl methanamine, benzyl methoxymethyl trimethylsilyl methyl amine, benzyl-methoxymethyl-trimethylsilanylmethyl-amine, n-benzyl-n-methoxymethyl-n-trimethylsilylmethylamine, n-benzyl-n-methoxymethyl trimethylsilylmethylamine, n-methoxymethyl phenyl-n-trimethylsilyl methyl methanamine, n-methoxymethyl-n-trimethylsilyl methyl phenyl, n-methoxymethyl-n-trimethylsilylmethyl-benzylamine
PubChem CID 353442
Nom de l’IUPAC N-(méthoxyméthyl)-1-phényl-N-(triméthylsilylméthyl)méthanamine
SOURIRES COCN(CC1=CC=CC=C1)C[Si](C)(C)C

Specifications

Nom ou substance chimique N-Benzyl-N-(methoxymethyl)-N-trimethylsilylmethylamine
Couleur Jaune
Point d’ébullition 78 °C
Quantité 5 g
Poids de formule 237.42
Pourcentage de pureté ≥98,0% (T)
Forme physique Liquide
TSCA Non
Recommended Storage Réfrigérateur