missing translation for 'onlineSavingsMsg'
Learn More

N-Benzoylsuccinimide, 97%, Thermo Scientific Chemicals

Catalog No. p-7027707
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2468006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2468006 Supplier Thermo Scientific Chemicals Supplier No. B2468006

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6343-27-7
Molecular Formula C11H9NO3
Molecular Weight (g/mol) 203.197
MDL Number MFCD00185021
InChI Key CLFHABXQJQAYEF-UHFFFAOYSA-N
Synonym n-benzoylsuccinimide, n-benzoyl succinimide, mira-b, acmc-20ak70, 1-benzoyl-2,5-pyrrolidinedione, 2,5-pyrrolidinedione,1-benzoyl, 1-phenylcarbonyl azolidine-2,5-dione
PubChem CID 232580
IUPAC Name 1-benzoylpyrrolidine-2,5-dione
SMILES C1CC(=O)N(C1=O)C(=O)C2=CC=CC=C2

Specifications

Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 179.18
Percent Purity 97%
Chemical Name or Material N-Benzoylsuccinimide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 653-674-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only