missing translation for 'onlineSavingsMsg'
Learn More

N-(Benzoyloxy)succinimide 98.0+%, TCI America™

Catalog No. b47855g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47855G 5 g
B478525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47855G Supplier TCI America Supplier No. B47855G
Only null left

Chemical Identifiers

CAS 23405-15-4
Molecular Formula C11H9NO4
Molecular Weight (g/mol) 219.196
MDL Number MFCD00078953
InChI Key BVUOEDOMUOJKOY-UHFFFAOYSA-N
Synonym n-benzoyloxy succinimide, bz-osu, n-succinimidyl benzoate, benzoic acid n-hydroxysuccinimide ester, 2,5-pyrrolidinedione, 1-benzoyloxy, 1-benzoyloxy-2,5-pyrrolidinedione, 2,5-dioxoazolidinyl benzoate, 1-phenylcarbonyl oxy pyrrolidine-2,5-dione, benzoyoxysuccinimide, benzoic acid succinimidyl ester
PubChem CID 716426
IUPAC Name (2,5-dioxopyrrolidin-1-yl) benzoate
SMILES C1CC(=O)N(C1=O)OC(=O)C2=CC=CC=C2

Specifications

Melting Point 137°C
Color White
Quantity 5 g
Formula Weight 219.20
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-(Benzoyloxy)succinimide
TSCA No
Recommended Storage Refrigerator