missing translation for 'onlineSavingsMsg'
Learn More

N-(Benzoyloxy)succinimide, 97%

Catalog No. p-7039392
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5862503 1 g
AAH5862506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5862503 Supplier Thermo Scientific Chemicals Supplier No. H5862503

CAS: 23405-15-4 | C11H9NO4 | 219.196 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23405-15-4
Molecular Formula C11H9NO4
Molecular Weight (g/mol) 219.196
MDL Number MFCD00078953
InChI Key BVUOEDOMUOJKOY-UHFFFAOYSA-N
Synonym n-benzoyloxy succinimide, bz-osu, n-succinimidyl benzoate, benzoic acid n-hydroxysuccinimide ester, 2,5-pyrrolidinedione, 1-benzoyloxy, 1-benzoyloxy-2,5-pyrrolidinedione, 2,5-dioxoazolidinyl benzoate, 1-phenylcarbonyl oxy pyrrolidine-2,5-dione, benzoyoxysuccinimide, benzoic acid succinimidyl ester
PubChem CID 716426
IUPAC Name (2,5-dioxopyrrolidin-1-yl) benzoate
SMILES C1CC(=O)N(C1=O)OC(=O)C2=CC=CC=C2

Specifications

Quantity 1 g
Formula Weight 219.19
Percent Purity 97%
Chemical Name or Material N-(Benzoyloxy)succinimide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only