missing translation for 'onlineSavingsMsg'
Learn More

N-Benzoyl-L-tyrosine Ethyl Ester 98.0+%, TCI America™

Catalog No. B11401G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11401G 1 g
B11405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B11401G Supplier TCI America Supplier No. B11401G
Only null left

Chemical Identifiers

CAS 3483-82-7
Molecular Formula C18H19NO4
Molecular Weight (g/mol) 313.35
MDL Number MFCD00002388
InChI Key SRLROPAFMUDDRC-XISACWJONA-N
Synonym n-benzoyl-l-tyrosine ethyl ester, bz-tyr-oet, ethyl n-benzoyl-l-tyrosinate, btee, ethyl benzoyltyrosinate, benzoyltyrosine ethyl ester, benzoyl-l-tyrosine ethyl ester, s-ethyl 2-benzamido-3-4-hydroxyphenyl propanoate, benzoyl-tyr-oet, l-tyrosine, n-benzoyl-, ethyl ester
PubChem CID 77033
IUPAC Name ethyl (2S)-3-(4-hydroxyphenyl)-2-(phenylformamido)propanoate
SMILES CCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)C1=CC=CC=C1

Specifications

Melting Point 122°C
Color White
Quantity 1 g
Formula Weight 313.35
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Benzoyl-L-tyrosine Ethyl Ester