missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzoyl-L-tyrosine ethyl ester, 98%
CAS: 3483-82-7 | C18H19NO4 | 313.35 g/mol
$71.03 - $250.70
Chemical Identifiers
| CAS | 3483-82-7 |
|---|---|
| Molecular Formula | C18H19NO4 |
| Molecular Weight (g/mol) | 313.35 |
| MDL Number | MFCD00002388 |
| InChI Key | SRLROPAFMUDDRC-XISACWJONA-N |
| Synonym | n-benzoyl-l-tyrosine ethyl ester, bz-tyr-oet, ethyl n-benzoyl-l-tyrosinate, btee, ethyl benzoyltyrosinate, benzoyltyrosine ethyl ester, benzoyl-l-tyrosine ethyl ester, s-ethyl 2-benzamido-3-4-hydroxyphenyl propanoate, benzoyl-tyr-oet, l-tyrosine, n-benzoyl-, ethyl ester |
| PubChem CID | 77033 |
| IUPAC Name | ethyl (2S)-2-benzamido-3-(4-hydroxyphenyl)propanoate |
| SMILES | CCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|
AAH6343406
|
Thermo Scientific Chemicals
H6343406 |
5 g |
Each for $71.03
|
|
|||||
|
AAH6343414
|
Thermo Scientific Chemicals
H6343414 |
25 g |
Each for $250.70
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 3483-82-7 | |
| 313.35 | |
| SRLROPAFMUDDRC-XISACWJONA-N | |
| 77033 | |
| CCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)C1=CC=CC=C1 |
| C18H19NO4 | |
| MFCD00002388 | |
| n-benzoyl-l-tyrosine ethyl ester, bz-tyr-oet, ethyl n-benzoyl-l-tyrosinate, btee, ethyl benzoyltyrosinate, benzoyltyrosine ethyl ester, benzoyl-l-tyrosine ethyl ester, s-ethyl 2-benzamido-3-4-hydroxyphenyl propanoate, benzoyl-tyr-oet, l-tyrosine, n-benzoyl-, ethyl ester | |
| ethyl (2S)-2-benzamido-3-(4-hydroxyphenyl)propanoate |
Specifications
| 118°C to 121°C | |
| C18H19NO4 | |
| 2223819 | |
| SRLROPAFMUDDRC-XISACWJONA-N | |
| ethyl (2S)-2-benzamido-3-(4-hydroxyphenyl)propanoate | |
| 313.35 | |
| 313.35 | |
| Solid |
| 3483-82-7 | |
| MFCD00002388 | |
| n-benzoyl-l-tyrosine ethyl ester, bz-tyr-oet, ethyl n-benzoyl-l-tyrosinate, btee, ethyl benzoyltyrosinate, benzoyltyrosine ethyl ester, benzoyl-l-tyrosine ethyl ester, s-ethyl 2-benzamido-3-4-hydroxyphenyl propanoate, benzoyl-tyr-oet, l-tyrosine, n-benzoyl-, ethyl ester | |
| CCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)C1=CC=CC=C1 | |
| 5 g | |
| 77033 | |
| 98% | |
| N-Benzoyl-L-tyrosine ethyl ester |
Safety and Handling
EINECSNumber : 222-469-2
TSCA : Yes
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only