missing translation for 'onlineSavingsMsg'
Learn More

N-Benzoyl-L-glutamic Acid 98.0+%, TCI America™

$31.68 - $150.98

Chemical Identifiers

CAS 6094-36-6
Molecular Formula C12H13NO5
Molecular Weight (g/mol) 251.238
MDL Number MFCD00014028
InChI Key LPJXPACOXRZCCP-VIFPVBQESA-N
Synonym bz-glu-oh, n-benzoyl-l-glutamic acid, benzoyl-l-glutamic acid, s-2-benzamidopentanedioic acid, n-benzoylglutamic acid, unii-3591d74mxp, 2s-2-phenylformamido pentanedioic acid, +-n-benzoylglutamic acid, n-4-benzoyl-l-glutamic acid, 2s-2-phenylcarbonylamino pentanedioic acid
PubChem CID 11345724
IUPAC Name (2S)-2-benzamidopentanedioic acid
SMILES C1=CC=C(C=C1)C(=O)NC(CCC(=O)O)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B02021G
View Documents
TCI America
B02021G
1 g N/A
Only null left
 
B020225G
View Documents
TCI America
B020225G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

6094-36-6
251.238
LPJXPACOXRZCCP-VIFPVBQESA-N
11345724
C1=CC=C(C=C1)C(=O)NC(CCC(=O)O)C(=O)O
C12H13NO5
MFCD00014028
bz-glu-oh, n-benzoyl-l-glutamic acid, benzoyl-l-glutamic acid, s-2-benzamidopentanedioic acid, n-benzoylglutamic acid, unii-3591d74mxp, 2s-2-phenylformamido pentanedioic acid, +-n-benzoylglutamic acid, n-4-benzoyl-l-glutamic acid, 2s-2-phenylcarbonylamino pentanedioic acid
(2S)-2-benzamidopentanedioic acid

Specifications

6094-36-6
White
MFCD00014028
bz-glu-oh, n-benzoyl-l-glutamic acid, benzoyl-l-glutamic acid, s-2-benzamidopentanedioic acid, n-benzoylglutamic acid, unii-3591d74mxp, 2s-2-phenylformamido pentanedioic acid, +-n-benzoylglutamic acid, n-4-benzoyl-l-glutamic acid, 2s-2-phenylcarbonylamino pentanedioic acid
C1=CC=C(C=C1)C(=O)NC(CCC(=O)O)C(=O)O
251.238
251.24
Crystalline Powder
140°C
C12H13NO5
1 g
LPJXPACOXRZCCP-VIFPVBQESA-N
(2S)-2-benzamidopentanedioic acid
11345724
≥98.0% (HPLC,T)
N-Benzoyl-L-glutamic Acid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'