missing translation for 'onlineSavingsMsg'
Learn More

N-Benzoyl-L-glutamic Acid 98.0+%, TCI America™

Catalog No. B020225G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B020225G 25 g
B02021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B020225G Supplier TCI America Supplier No. B020225G
Only null left

Chemical Identifiers

CAS 6094-36-6
Molecular Formula C12H13NO5
Molecular Weight (g/mol) 251.238
MDL Number MFCD00014028
InChI Key LPJXPACOXRZCCP-VIFPVBQESA-N
Synonym bz-glu-oh, n-benzoyl-l-glutamic acid, benzoyl-l-glutamic acid, s-2-benzamidopentanedioic acid, n-benzoylglutamic acid, unii-3591d74mxp, 2s-2-phenylformamido pentanedioic acid, +-n-benzoylglutamic acid, n-4-benzoyl-l-glutamic acid, 2s-2-phenylcarbonylamino pentanedioic acid
PubChem CID 11345724
IUPAC Name (2S)-2-benzamidopentanedioic acid
SMILES C1=CC=C(C=C1)C(=O)NC(CCC(=O)O)C(=O)O

Specifications

Melting Point 140°C
Color White
Quantity 25 g
Formula Weight 251.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Benzoyl-L-glutamic Acid