missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-alpha-Benzoyl-DL-arginine-4-nitroanilide hydrochloride, 98%

Catalog No. AC227740010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC227740010 1 g Glass Bottle
AC227740050 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC227740010 Supplier Thermo Scientific Chemicals Supplier No. 227740010

CAS: 911-77-3 | C19H23ClN6O4 | 434.881 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 911-77-3
Molecular Formula C19H23ClN6O4
Molecular Weight (g/mol) 434.881
MDL Number MFCD00012846
InChI Key DEOKFPFLXFNAON-UHFFFAOYSA-N
Synonym bapna, n-5-guanidino-1-4-nitrophenyl amino-1-oxopentan-2-yl benzamide hydrochloride, n-benzoyl-dl-arginine-4-nitroanilide hydrochloride, n-alpha-benzoyl-dl-arginine-4-nitroanilide hydrochloride, nalpha-benzoyl-dl-arginine 4-nitroanilide hydrochloride, bani, 5-carbamimidamido-n-4-nitrophenyl-2-phenylformamido pentanamide hydrochloride, l-bapna, dl-bapa, bz-dl-arg-pna hcl
PubChem CID 2724371
IUPAC Name N-[5-(diaminomethylideneamino)-1-(4-nitroanilino)-1-oxopentan-2-yl]benzamide;hydrochloride
SMILES C1=CC=C(C=C1)C(=O)NC(CCCN=C(N)N)C(=O)NC2=CC=C(C=C2)[N+](=O)[O-].Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 434.89
Percent Purity ≥97.5% (HPLC)
Physical Form Powder
Chemical Name or Material N-α-Benzoyl-DL-arginine-4-nitroanilide hydrochloride
Show More Show Less

RUO – Research Use Only