missing translation for 'onlineSavingsMsg'
Learn More

N-Aminosuccinimide Hydrochloride 98.0+%, TCI America™

Catalog No. p-7117595
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A18191G 1 g
A18195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A18191G Supplier TCI America Supplier No. A18191G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108906-15-6
Molecular Formula C4H7ClN2O2
Molecular Weight (g/mol) 150.562
InChI Key SRKWEKIPNNNHSG-UHFFFAOYSA-N
PubChem CID 12480332
IUPAC Name 1-aminopyrrolidine-2,5-dione;hydrochloride
SMILES C1CC(=O)N(C1=O)N.Cl

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 150.56
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Aminosuccinimide Hydrochloride
TSCA No
Recommended Storage Refrigerator