missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Aminophthalimide, 94%
Description
It is employed in the aziridination of chiral N-enoyl sultams. It is also used in the synthesis of n-phthalimidoaziridines
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed in the aziridination of chiral N-enoyl sultams. It is also used in the synthesis of n-phthalimidoaziridines
Solubility
Solubility in hot methanol almost transparent.
Notes
Store away from oxidizing agents, strong bases and heat. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
It is employed in the aziridination of chiral N-enoyl sultams. It is also used in the synthesis of n-phthalimidoaziridines
Solubility
Solubility in hot methanol almost transparent.
Notes
Store away from oxidizing agents, strong bases and heat. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
Specifications
Specifications
| Melting Point | ∼195°C (decomposition) |
| Quantity | 5 g |
| Beilstein | 383756 |
| Formula Weight | 162.15 |
| Percent Purity | 94% |
| Chemical Name or Material | N-Aminophthalimide |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 217-505-9 |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only