missing translation for 'onlineSavingsMsg'
Learn More

N-Aminophthalimide, 94%

Catalog No. AAA1991418
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1991418 50 g
AAA1991406 5 g
AAA1991409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1991418 Supplier Thermo Scientific Chemicals Supplier No. A1991418
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1875-48-5 | C8H6N2O2 | 162.148 g/mol

It is employed in the aziridination of chiral N-enoyl sultams. It is also used in the synthesis of n-phthalimidoaziridines

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the aziridination of chiral N-enoyl sultams. It is also used in the synthesis of n-phthalimidoaziridines

Solubility
Solubility in hot methanol almost transparent.

Notes
Store away from oxidizing agents, strong bases and heat. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.

Chemical Identifiers

CAS 1875-48-5
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD00005895
InChI Key KSILMCDYDAKOJD-UHFFFAOYSA-N
Synonym n-aminophthalimide, 2-aminoisoindoline-1,3-dione, 2-amino-1h-isoindole-1,3 2h-dione, phthalimide, n-amino, hydrazine, 1,1-phthaloyl, phthalhydrazide van, 1h-isoindole-1,3 2h-dione, 2-amino, n-aminophthalamide, phthalocyclohydrazide van, ccris 5349
PubChem CID 74645
IUPAC Name 2-aminoisoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)N

Specifications

Melting Point ∼195°C (decomposition)
Quantity 50 g
Beilstein 383756
Formula Weight 162.15
Percent Purity 94%
Chemical Name or Material N-Aminophthalimide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-505-9
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only