missing translation for 'onlineSavingsMsg'
Learn More

N-Aminomorpholine, 95%

CAS: 4319-49-7 | C4H10N2O | 102.14 g/mol

$134.69 - $134.69

Chemical Identifiers

CAS 4319-49-7
Molecular Formula C4H10N2O
Molecular Weight (g/mol) 102.14
MDL Number MFCD00006174
InChI Key MKQLBNJQQZRQJU-UHFFFAOYSA-N
Synonym 4-aminomorpholine, n-aminomorpholine, 4-morpholinamine, morpholin-4-ylamine, morpholine, 4-amino, 4-morpholinamine 9ci, morpholine-4-ylamine, n-amino morpholine, n-amino-morpholine, 4-amino morpholine
PubChem CID 20315
IUPAC Name morpholin-4-amine
SMILES C1COCCN1N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC104110050
SDS View Documents
Thermo Scientific Chemicals
104110050
5 g Glass bottle
Each for $134.69
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4319-49-7
102.14
MKQLBNJQQZRQJU-UHFFFAOYSA-N
20315
C1COCCN1N
C4H10N2O
MFCD00006174
4-aminomorpholine, n-aminomorpholine, 4-morpholinamine, morpholin-4-ylamine, morpholine, 4-amino, 4-morpholinamine 9ci, morpholine-4-ylamine, n-amino morpholine, n-amino-morpholine, 4-amino morpholine
morpholin-4-amine

Specifications

4319-49-7
100.0
1.0500g/mL
58°C
96% min. (GC)
C4H10N2O
MFCD00006174
27, 8
4-aminomorpholine, n-aminomorpholine, 4-morpholinamine, morpholin-4-ylamine, morpholine, 4-amino, 4-morpholinamine 9ci, morpholine-4-ylamine, n-amino morpholine, n-amino-morpholine, 4-amino morpholine
MKQLBNJQQZRQJU-UHFFFAOYSA-N
morpholin-4-amine
20315
95%
N-Aminomorpholine, 97%
96.0
Colorless to Yellow
168.0°C
Authentic
Glass bottle
1.4765 to 1.4785
5 g
1.05
Solubility in water: miscible.
C1COCCN1N
102.14
102.14
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 224-347-4

RTECSNumber : QD7300000

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only