missing translation for 'onlineSavingsMsg'
Learn More

N(alpha)-Boc-L-arginine, 98%

Catalog No. p-7035660
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5337903 1 g
AAH5337906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5337903 Supplier Thermo Scientific Chemicals Supplier No. H5337903

CAS: 13726-76-6 | C11H22N4O4 | 274.321 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13726-76-6
Molecular Formula C11H22N4O4
Molecular Weight (g/mol) 274.321
MDL Number MFCD00042632
InChI Key HSQIYOPBCOPMSS-ZETCQYMHSA-N
Synonym boc-arg-oh, boc-arginine, s-2-tert-butoxycarbonyl amino-5-guanidinopentanoic acid, boc-l-arg-oh, n2-tert-butoxycarbonyl-l-arginine, n-boc-l-arginine, na-tert-butoxycarbonyl-l-arginine, n-t-boc-l-arginine, n alpha-boc-l-arginine, n∼2∼-tert-butoxycarbonyl-l-arginine
PubChem CID 114667
IUPAC Name (2S)-5-(diaminomethylideneamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
SMILES CC(C)(C)OC(=O)NC(CCCN=C(N)N)C(=O)O

Specifications

Flash Point 113°C (235°F)
Quantity 1 g
Beilstein 2419628
Optical Rotation −6.5° (c=1 in Acetic Acid)
Formula Weight 274.32
Percent Purity 98%
Chemical Name or Material N(alpha)-Boc-L-arginine
EINECSNumber 237-295-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only