missing translation for 'onlineSavingsMsg'
Learn More

N(alpha)-Boc-L-2,3-diaminopropionic acid, 97%

Catalog No. AAH2691903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2691903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2691903 Supplier Thermo Scientific Chemicals Supplier No. H2691903
Only null left

CAS: 73259-81-1 | C8H16N2O4 | 204.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 73259-81-1
Molecular Formula C8H16N2O4
Molecular Weight (g/mol) 204.23
MDL Number MFCD00236843
InChI Key KRJLRVZLNABMAT-YFKPBYRVSA-N
Synonym boc-dap-oh, 3-amino-boc-l-alanine, boc-l-2,3-diaminopropionic acid, s-3-amino-2-tert-butoxycarbonyl amino propanoic acid, n-boc-beta-amino-ala-oh, boc-dpr-oh, 2s-3-amino-2-tert-butoxy carbonyl amino propanoic acid, 3-amino-n-boc-l-alanine, n-boc-l-2,3-diaminopropanoic acid, nalpha-boc-2,3-diaminopropionic acid
PubChem CID 2755946
IUPAC Name (2S)-3-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](C[NH3+])C([O-])=O

Specifications

Melting Point ∽210°C (decomposition)
Quantity 1 g
Optical Rotation −14° (c=1 in Water)
Formula Weight 204.23
Percent Purity 97%
Chemical Name or Material N(alpha)-Boc-L-2,3-diaminopropionic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only