missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N(alpha)-Boc-D-histidine, 98+%

Catalog No. p-7044942
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL08810MD 250 mg
AAL0881003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL08810MD Supplier Thermo Scientific Chemicals Supplier No. L08810MD
Only null left

CAS: 50654-94-9 | C11H17N3O4 | 255.274 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50654-94-9
Molecular Formula C11H17N3O4
Molecular Weight (g/mol) 255.274
MDL Number MFCD00037851
InChI Key AYMLQYFMYHISQO-MRVPVSSYSA-N
Synonym boc-d-his-oh, boc-d-histidine, n-a-boc-d-histidine, d-histidine, n-1,1-dimethylethoxy carbonyl, n-alpha-t-butyloxycarbonyl-d-histidine, r-2-tert-butoxycarbonyl amino-3-1h-imidazol-4-yl propanoic acid, r-2-tert-butoxycarbonyl amino-3-1h-imidazol-4-yl propionic acid, n-alpha-boc-d-histidine, 2r-2-tert-butoxycarbonyl amino-3-1h-imidazol-4-yl propanoic acid, n-boc-d-histidine
PubChem CID 2724761
IUPAC Name (2R)-3-(1H-imidazol-5-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CN=CN1)C(=O)O

Specifications

Melting Point ∽197°C (decomposition)
Beilstein 4196575
Quantity 250 mg
Formula Weight 255.27
Optical Rotation −26° (c=1 in Methanol)
Percent Purity ≥98%
Chemical Name or Material N(alpha)-Boc-D-histidine
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only