missing translation for 'onlineSavingsMsg'
Learn More

N(alpha)-Benzyloxycarbonyl-L-tryptophan, 98+%

Catalog No. AAL0753514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0753514 25 g
AAL0753506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0753514 Supplier Thermo Scientific Chemicals Supplier No. L0753514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7432-21-5 | C19H18N2O4 | 338.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7432-21-5
Molecular Formula C19H18N2O4
Molecular Weight (g/mol) 338.36
MDL Number MFCD00065700
InChI Key AHYFYYVVAXRMKB-KRWDZBQOSA-N
Synonym z-trp-oh, n-cbz-l-tryptophan, z-l-trp-oh, benzyloxycarbonyltryptophan, l-tryptophan, n-phenylmethoxy carbonyl, n-carbobenzoxyltryptophan, n-carbobenzoxy-l-tryptophan, cbz-l-trp-oh, n-benzyloxycarbonyl-l-tryptophan, nalpha-carbobenzyloxy-l-tryptophan
PubChem CID 101176
IUPAC Name (2S)-3-(1H-indol-3-yl)-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES OC(=O)[C@H](CC1=CNC2=CC=CC=C12)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 121°C to 126°C
Beilstein 96743
Quantity 25 g
Odor Odorless
Formula Weight 338.37
Optical Rotation +3.1° (c=3 in Acetic Acid)
Percent Purity ≥98%
Chemical Name or Material N(alpha)-Benzyloxycarbonyl-L-tryptophan
EINECSNumber 231-074-4
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only