missing translation for 'onlineSavingsMsg'
Learn More

N(alpha)-Benzyloxycarbonyl-L-lysine, 98+%

Catalog No. p-7044892
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0862903 1 g
AAL0862906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0862903 Supplier Thermo Scientific Chemicals Supplier No. L0862903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2212-75-1 | C14H20N2O4 | 280.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2212-75-1
Molecular Formula C14H20N2O4
Molecular Weight (g/mol) 280.32
MDL Number MFCD00038204
InChI Key OJTJKAUNOLVMDX-LBPRGKRZSA-N
Synonym z-lys-oh, nalpha-cbz-l-lysine, n-alpha-cbz-l-lysine, cbz-lys-oh, n-alpha-carbobenzoxy-l-lysine, s-6-amino-2-benzyloxy carbonyl amino hexanoic acid, z-lysine, cbz-l-lysine, n-alpha-benzyloxycarbonyl-l-lysine, lysine, n2-phenylmethoxy carbonyl
PubChem CID 75172
IUPAC Name (2S)-6-amino-2-(phenylmethoxycarbonylamino)hexanoic acid
SMILES NCCCC[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point ∽237°C (decomposition)
Beilstein 2153826
Quantity 1 g
Formula Weight 280.32
Optical Rotation −12° (c=2 in 0.2N HCl)
Percent Purity ≥98%
Chemical Name or Material N(alpha)-Benzyloxycarbonyl-L-lysine
EINECSNumber 218-662-6
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only