missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-alpha-Benzoyl-L-arginine p-nitroanilide hydrochloride

Catalog No. AAJ64915MC
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64915MC 100 mg
AAJ64915MD 250 mg
AAJ64915ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ64915MC Supplier Thermo Scientific Chemicals Supplier No. J64915MC

CAS: 21653-40-7 | C19H23ClN6O4 | 434.881 g/mol

N-alpha-Benzoyl-L-arginine p-nitroanilide hydrochloride is used as a chromogenic substrate for trypsin and other proteolytic enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-alpha-Benzoyl-L-arginine p-nitroanilide hydrochloride is used as a chromogenic substrate for trypsin and other proteolytic enzymes.

Solubility
Soluble in DMSO at 50mg/ml

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 21653-40-7
Molecular Formula C19H23ClN6O4
Molecular Weight (g/mol) 434.881
MDL Number MFCD00063682
InChI Key DEOKFPFLXFNAON-NTISSMGPSA-N
Synonym bz-arg-pna.hcl, s-n-5-guanidino-1-4-nitrophenyl amino-1-oxopentan-2-yl benzamide hydrochloride, nalpha-benzoyl-l-arginine 4-nitroanilide hydrochloride, bani, s-5-amidinoamino-2-benzoylamino-n-4-nitrophenyl valeramide monohydrochloride, l-bapna, 2s-5-carbamimidamido-n-4-nitrophenyl-2-phenylformamido pentanamide hydrochloride, bz-arg-pna-hcl, bz-arg-pna hcl, bz-arg-pna . hcl
PubChem CID 16219022
IUPAC Name N-[(2S)-5-(diaminomethylideneamino)-1-(4-nitroanilino)-1-oxopentan-2-yl]benzamide;hydrochloride
SMILES C1=CC=C(C=C1)C(=O)NC(CCCN=C(N)N)C(=O)NC2=CC=C(C=C2)[N+](=O)[O-].Cl

Specifications

Melting Point 223°C
Color White to Yellow
Beilstein 4081878
Solubility Information Soluble in DMSO at 50mg/ml
Quantity 100 mg
Formula Weight 434.9
Physical Form Powder
Chemical Name or Material N-alpha-Benzoyl-L-arginine p-nitroanilide hydrochloride
EINECSNumber 244-505-6
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only