missing translation for 'onlineSavingsMsg'
Learn More

N-alpha-Benzoyl-L-arginine ethyl ester hydrochloride, 98+%

Catalog No. AAA1818106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1818106 5 g
AAA1818103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1818106 Supplier Thermo Scientific Chemicals Supplier No. A1818106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2645-08-1 | C16H17ClN4O2S | 364.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2645-08-1
MDL Number MFCD00012579

Specifications

Melting Point 127°C to 131°C
Molecular Formula C15H23ClN4O3
Beilstein 3781694
Synonym bz-arg-oet.hcl, n-alpha-benzoyl-l-arginine ethyl ester hydrochloride, ethyl n-benzoyl-l-argininate hydrochloride, baee, nalpha-benzoyl-l-arginine ethyl ester hydrochloride, l-baee, l-arginine, n2-benzoyl-, ethyl ester, monohydrochloride, n-benzoyl-l-arginine ethyl ester hydrochloride, ethyl n2-benzoyl-l-argininate monohydrochloride, ethyl 2s-5-carbamimidamido-2-phenylformamido pentanoate hydrochloride
InChI Key HIXDELXKSSLIKB-UHFFFAOYNA-N
SMILES CCOC(=O)C(CCCN=C(N)N)NC(=O)C1=CC=CC=C1.Cl
IUPAC Name ethyl (2S)-2-benzamido-5-(diaminomethylideneamino)pentanoate;hydrochloride
Quantity 5 g
Molecular Weight (g/mol) 342.82
PubChem CID 2723604
Formula Weight 342.84
Optical Rotation −17° (c=2, Water)
Percent Purity ≥98%
Sensitivity Hygroscopic
Chemical Name or Material N-alpha-Benzoyl-L-arginine ethyl ester hydrochloride
Show More Show Less
EINECSNumber 220-157-0
TSCA No
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only