missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N(alpha)-Acetyl-L-lysine, 99%

Catalog No. AAJ6508303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6508303 1 g
AAJ6508306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6508303 Supplier Thermo Scientific Chemicals Supplier No. J6508303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1946-82-3 | C8H16N2O3 | 188.23 g/mol

Nα-Acetyl-L-lysine is a biologically available N-terminal capped form of the proteinogenic α amino acid L-lysine. Nα-Acetyl-L-lysine is a substrate used to study and characterize lysyl oxidize(s).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nα-Acetyl-L-lysine is a biologically available N-terminal capped form of the proteinogenic α amino acid L-lysine. Nα-Acetyl-L-lysine is a substrate used to study and characterize lysyl oxidize(s).

Solubility
Soluble in water (miscible), and 80% acetic acid (50 mg/ml--clear colorless solution)).

Notes
Protect from heat. Store away from oxidizing agents.

Chemical Identifiers

CAS 1946-82-3
Molecular Formula C8H16N2O3
Molecular Weight (g/mol) 188.23
MDL Number MFCD00008233
InChI Key VEYYWZRYIYDQJM-JLDDOWRYNA-N
Synonym ac-lys-oh, n-alpha-acetyl-l-lysine, n alpha-acetyllysine, nalpha-acetyl-l-lysine, n-acetyl-l-lysine, n2-acetyl-l-lysine, s-2-acetamido-6-aminohexanoic acid, n alpha-acetyl-l-lysine, n 2-acetyl-l-lysine, acetyl-l-lysine
PubChem CID 92907
ChEBI CHEBI:35704
IUPAC Name (2S)-2-acetamido-6-aminohexanoic acid
SMILES CC(=O)N[C@@H](CCCCN)C(O)=O

Specifications

Melting Point 256°C to 258°C (decomposition)
Color White
Solubility Information Soluble in water (miscible),and 80% acetic acid (50mg/ml--clear colorless solution)).
Quantity 1 g
Formula Weight 188.2
Percent Purity 99%
Physical Form Powder
Chemical Name or Material N(alpha)-Acetyl-L-lysine
EINECSNumber 217-747-5
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only