missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[N-[alpha-[2-(Dibutylglycinamido)phenyl]benzylidene]glycinato]nickel 95.0+%, TCI America™
$50.05 - $313.80
Chemical Identifiers
| CAS | 847654-16-4 |
|---|---|
| Molecular Formula | C25H31N3NiO3 |
| Molecular Weight (g/mol) | 480.23 |
| MDL Number | MFCD10566890 |
| InChI Key | KZZGXIZCIUYKSB-UHFFFAOYSA-L |
| PubChem CID | 53384304 |
| IUPAC Name | nickel(2+) 2-{[(2-{[2-(dibutylamino)-1-oxidoethylidene]amino}phenyl)(phenyl)methylidene]amino}acetate |
| SMILES | [Ni++].CCCCN(CCCC)CC([O-])=NC1=CC=CC=C1C(=NCC([O-])=O)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D3543100MG
|
TCI America
D3543100MG |
100 mg |
Each for $50.05
|
|
|||||
|
D35431G
|
TCI America
D35431G |
1 g |
Each for $313.80
|
|
|||||
Chemical Identifiers
| 847654-16-4 | |
| 480.23 | |
| KZZGXIZCIUYKSB-UHFFFAOYSA-L | |
| nickel(2+) 2-{[(2-{[2-(dibutylamino)-1-oxidoethylidene]amino}phenyl)(phenyl)methylidene]amino}acetate |
| C25H31N3NiO3 | |
| MFCD10566890 | |
| 53384304 | |
| [Ni++].CCCCN(CCCC)CC([O-])=NC1=CC=CC=C1C(=NCC([O-])=O)C1=CC=CC=C1 |
Specifications
| 847654-16-4 | |
| MFCD10566890 | |
| KZZGXIZCIUYKSB-UHFFFAOYSA-L | |
| nickel(2+) 2-{[(2-{[2-(dibutylamino)-1-oxidoethylidene]amino}phenyl)(phenyl)methylidene]amino}acetate | |
| 53384304 | |
| ≥95.0% (T) | |
| [N-[alpha-[2-(Dibutylglycinamido)phenyl]benzylidene]glycinato]nickel |
| C25H31N3NiO3 | |
| 100 mg | |
| [Ni++].CCCCN(CCCC)CC([O-])=NC1=CC=CC=C1C(=NCC([O-])=O)C1=CC=CC=C1 | |
| 480.23 | |
| 480.23 | |
| Crystalline Powder |
Safety and Handling
TSCA : No