missing translation for 'onlineSavingsMsg'
Learn More

N-Allylmethylamine, 96%

Catalog No. AAH2755103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2755103 1 g
AAH2755106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2755103 Supplier Thermo Scientific Chemicals Supplier No. H2755103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 627-37-2 | C4H9N | 71.123 g/mol

N-Allylmethylamine is used in the production of N-allyl-alfa,alfa-dichloro-N-methylacetamide. It is used as a functional monomer to coordinate with Cd(II)/Zn(II) ions to get the poly methyl methacrylate -butyl methacrylate - N-allylmethylamine (PMBA) films, which is useful as building blocks to fabricate a photo-catalytic cell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Allylmethylamine is used in the production of N-allyl-alfa,alfa-dichloro-N-methylacetamide. It is used as a functional monomer to coordinate with Cd(II)/Zn(II) ions to get the poly methyl methacrylate -butyl methacrylate - N-allylmethylamine (PMBA) films, which is useful as building blocks to fabricate a photo-catalytic cell.

Solubility
Miscible with water.

Notes
Store in a cool place. Incompatible with oxidizing agents, strong acids and bases.

Chemical Identifiers

CAS 627-37-2
Molecular Formula C4H9N
Molecular Weight (g/mol) 71.123
MDL Number MFCD00008641
InChI Key IOXXVNYDGIXMIP-UHFFFAOYSA-N
Synonym n-allylmethylamine, n-methylallylamine, 2-propen-1-amine, n-methyl, allylmethylamine, methylallylamine, n-methyl-2-propen-1-amine, n-allyl-n-methylamine, methyl prop-2-en-1-yl amine, allylamine, n-methyl-7ci,8ci, 2-propen-1-amine, n-methyl-9ci
PubChem CID 69391
IUPAC Name N-methylprop-2-en-1-amine
SMILES CNCC=C

Specifications

Density 0.748
Boiling Point 61°C to 62°C
Flash Point −8°C (−18°F)
Refractive Index 1.412
Quantity 1 g
UN Number UN2733
Solubility Information Miscible with water.
Formula Weight 71.12
Percent Purity 96%
Chemical Name or Material N-Allylmethylamine
Hazard Category H225-H301+H311+H331-H314-H335
Hazard Statement GHS H Statement
H225-H301-H311-H314-H318-H332
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 210-996-0
RTECSNumber UC6632500
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only