missing translation for 'onlineSavingsMsg'
Learn More

N-Allylaniline 99.0+%, TCI America™

Catalog No. p-7117105
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Catalog No. Quantity
A088025ML 25 mL
A0880100ML 100 mL
A0880500ML 500 mL
3 options

Catalog No. A088025ML

Supplier: TCI America A088025ML

Only null left

Chemical Identifiers

CAS 589-09-3
Molecular Formula C9H11N
Molecular Weight (g/mol) 133.19
MDL Number MFCD00008638
InChI Key LQFLWKPCQITJIH-UHFFFAOYSA-N
Synonym n-allylaniline, n-allyl-aniline, benzenamine, n-2-propenyl, unii-q7s639gwxg, n-prop-2-en-1-ylaniline, q7s639gwxg, phenylprop-2-enylamine, nyl, allylphenylamine, allyl-phenyl-amine
PubChem CID 68525
IUPAC Name N-(prop-2-en-1-yl)aniline
SMILES C=CCNC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 220°C
Quantity 25 mL
Formula Weight 133.19
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Allylaniline