missing translation for 'onlineSavingsMsg'
Learn More

N-Allylaniline 99.0+%, TCI America™

Catalog No. A088025ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A088025ML 25 mL
A0880100ML 100 mL
A0880500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A088025ML Supplier TCI America Supplier No. A088025ML
Only null left

Chemical Identifiers

CAS 589-09-3
Molecular Formula C9H11N
Molecular Weight (g/mol) 133.19
MDL Number MFCD00008638
InChI Key LQFLWKPCQITJIH-UHFFFAOYSA-N
Synonym n-allylaniline, n-allyl-aniline, benzenamine, n-2-propenyl, unii-q7s639gwxg, n-prop-2-en-1-ylaniline, q7s639gwxg, phenylprop-2-enylamine, nyl, allylphenylamine, allyl-phenyl-amine
PubChem CID 68525
IUPAC Name N-(prop-2-en-1-yl)aniline
SMILES C=CCNC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 220°C
Quantity 25 mL
Formula Weight 133.19
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Allylaniline