missing translation for 'onlineSavingsMsg'
Learn More

N-Allylaniline, 95%, Thermo Scientific™

Catalog No. p-52538
Change view
Click to view available options
Quantity:
5g
25g
100g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC102870050 5g
AC102870250 25g
AC102871000 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC102870050 Supplier Thermo Scientific Chemicals Supplier No. 102870050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 589-09-3
Molecular Formula C9H11N
Molecular Weight (g/mol) 133.19
MDL Number MFCD00008638
InChI Key LQFLWKPCQITJIH-UHFFFAOYSA-N
Synonym n-allylaniline, n-allyl-aniline, benzenamine, n-2-propenyl, unii-q7s639gwxg, n-prop-2-en-1-ylaniline, q7s639gwxg, phenylprop-2-enylamine, nyl, allylphenylamine, allyl-phenyl-amine
PubChem CID 68525
IUPAC Name N-prop-2-enylaniline
SMILES C=CCNC1=CC=CC=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Density 0.9820g/mL
Boiling Point 218°C to 220°C
Flash Point 89°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Refractive Index 1.5620 to 1.564
Linear Formula C6H5NHCH2CH=CH2
Quantity 5g
Beilstein 12, 170
Specific Gravity 0.982
Formula Weight 133.19
Percent Purity 95%
Chemical Name or Material N-Allylaniline, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-633-9

RUO – Research Use Only