missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetylimidazole 98.0+%, TCI America™
$162.17 - $162.17
Chemical Identifiers
| CAS | 2466-76-4 |
|---|---|
| Molecular Formula | C5H6N2O |
| Molecular Weight (g/mol) | 110.12 |
| MDL Number | MFCD00005287 |
| InChI Key | VIHYIVKEECZGOU-UHFFFAOYSA-N |
| Synonym | 1-acetylimidazole, n-acetylimidazole, 1-1h-imidazol-1-yl ethanone, 1h-imidazole, 1-acetyl, acetylimidazole, 1-acetyl-1h-imidazole, imidazole, 1-acetyl, 3-acetylimidazole, unii-zmp8x1y11g, ccris 6534 |
| PubChem CID | 17174 |
| ChEBI | CHEBI:16984 |
| IUPAC Name | 1-(1H-imidazol-1-yl)ethan-1-one |
| SMILES | CC(=O)N1C=CN=C1 |
Chemical Identifiers
| 2466-76-4 | |
| 110.12 | |
| VIHYIVKEECZGOU-UHFFFAOYSA-N | |
| 17174 | |
| 1-(1H-imidazol-1-yl)ethan-1-one |
| C5H6N2O | |
| MFCD00005287 | |
| 1-acetylimidazole, n-acetylimidazole, 1-1h-imidazol-1-yl ethanone, 1h-imidazole, 1-acetyl, acetylimidazole, 1-acetyl-1h-imidazole, imidazole, 1-acetyl, 3-acetylimidazole, unii-zmp8x1y11g, ccris 6534 | |
| CHEBI:16984 | |
| CC(=O)N1C=CN=C1 |
Specifications
| 2466-76-4 | |
| White-Yellow | |
| MFCD00005287 | |
| 1-acetylimidazole, n-acetylimidazole, 1-1h-imidazol-1-yl ethanone, 1h-imidazole, 1-acetyl, acetylimidazole, 1-acetyl-1h-imidazole, imidazole, 1-acetyl, 3-acetylimidazole, unii-zmp8x1y11g, ccris 6534 | |
| CC(=O)N1C=CN=C1 | |
| 110.12 | |
| CHEBI:16984 | |
| ≥98.0% (GC,T) | |
| N-Acetylimidazole |
| 102°C | |
| C5H6N2O | |
| 25 g | |
| VIHYIVKEECZGOU-UHFFFAOYSA-N | |
| 1-(1H-imidazol-1-yl)ethan-1-one | |
| 17174 | |
| 110.12 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : NI3400000
TSCA : Yes