missing translation for 'onlineSavingsMsg'
Learn More

N-Acetylimidazole 98.0+%, TCI America™

Catalog No. A069425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A069425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A069425G Supplier TCI America Supplier No. A069425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2466-76-4
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD00005287
InChI Key VIHYIVKEECZGOU-UHFFFAOYSA-N
Synonym 1-acetylimidazole, n-acetylimidazole, 1-1h-imidazol-1-yl ethanone, 1h-imidazole, 1-acetyl, acetylimidazole, 1-acetyl-1h-imidazole, imidazole, 1-acetyl, 3-acetylimidazole, unii-zmp8x1y11g, ccris 6534
PubChem CID 17174
ChEBI CHEBI:16984
IUPAC Name 1-(1H-imidazol-1-yl)ethan-1-one
SMILES CC(=O)N1C=CN=C1

Specifications

Melting Point 102°C
Color White-Yellow
Quantity 25 g
Formula Weight 110.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetylimidazole
RTECSNumber NI3400000
TSCA Yes