missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetylglycine 99.0+%, TCI America™
$37.29 - $169.63
Chemical Identifiers
| CAS | 543-24-8 |
|---|---|
| Molecular Formula | C4H7NO3 |
| Molecular Weight (g/mol) | 117.104 |
| MDL Number | MFCD00004275 |
| InChI Key | OKJIRPAQVSHGFK-UHFFFAOYSA-N |
| Synonym | n-acetylglycine, aceturic acid, acetamidoacetic acid, acetylglycine, ac-gly-oh, glycine, n-acetyl, acetylaminoacetic acid, acetylamino-acetic acid, acetylglycocoll, ethanoylaminoethanoic acid |
| PubChem CID | 10972 |
| ChEBI | CHEBI:40410 |
| IUPAC Name | 2-acetamidoacetic acid |
| SMILES | CC(=O)NCC(=O)O |
Chemical Identifiers
| 543-24-8 | |
| 117.104 | |
| OKJIRPAQVSHGFK-UHFFFAOYSA-N | |
| 10972 | |
| 2-acetamidoacetic acid |
| C4H7NO3 | |
| MFCD00004275 | |
| n-acetylglycine, aceturic acid, acetamidoacetic acid, acetylglycine, ac-gly-oh, glycine, n-acetyl, acetylaminoacetic acid, acetylamino-acetic acid, acetylglycocoll, ethanoylaminoethanoic acid | |
| CHEBI:40410 | |
| CC(=O)NCC(=O)O |
Specifications
| 543-24-8 | |
| White | |
| MFCD00004275 | |
| n-acetylglycine, aceturic acid, acetamidoacetic acid, acetylglycine, ac-gly-oh, glycine, n-acetyl, acetylaminoacetic acid, acetylamino-acetic acid, acetylglycocoll, ethanoylaminoethanoic acid | |
| CC(=O)NCC(=O)O | |
| 117.104 | |
| CHEBI:40410 | |
| ≥99.0% (T) | |
| N-Acetylglycine |
| 207°C | |
| C4H7NO3 | |
| 25 g | |
| OKJIRPAQVSHGFK-UHFFFAOYSA-N | |
| 2-acetamidoacetic acid | |
| 10972 | |
| 117.10 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-1204
TSCA : Yes