missing translation for 'onlineSavingsMsg'
Learn More

N-Acetylglycine 99.0+%, TCI America™

Catalog No. A0093500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0093500G 500 g
A009325G 25 g
A0093100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A0093500G Supplier TCI America Supplier No. A0093500G

Chemical Identifiers

CAS 543-24-8
Molecular Formula C4H7NO3
Molecular Weight (g/mol) 117.104
MDL Number MFCD00004275
InChI Key OKJIRPAQVSHGFK-UHFFFAOYSA-N
Synonym n-acetylglycine, aceturic acid, acetamidoacetic acid, acetylglycine, ac-gly-oh, glycine, n-acetyl, acetylaminoacetic acid, acetylamino-acetic acid, acetylglycocoll, ethanoylaminoethanoic acid
PubChem CID 10972
ChEBI CHEBI:40410
IUPAC Name 2-acetamidoacetic acid
SMILES CC(=O)NCC(=O)O

Specifications

Melting Point 207°C
Color White
Quantity 500 g
Formula Weight 117.10
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetylglycine
EINECSNumber (2)-1204
TSCA Yes