missing translation for 'onlineSavingsMsg'
Learn More

N-Acetylglycine Ethyl Ester 98.0+%, TCI America™

Catalog No. p-7116711
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A00941G 1 g
A009425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A00941G Supplier TCI America Supplier No. A00941G

Chemical Identifiers

CAS 1906-82-7
Molecular Formula C6H11NO3
Molecular Weight (g/mol) 145.158
MDL Number MFCD00009173
InChI Key AMBDTBHJFINMSE-UHFFFAOYSA-N
Synonym ethyl acetamidoacetate, ac-gly-oet, n-acetylglycine ethyl ester, ethyl n-acetylglycinate, ethylaceturate, ethyl acetaminoacetate, ethyl 2-acetylamino acetate, glycine, n-acetyl-, ethyl ester, acetylglycine ethyl ester, acetyl-glycine ethyl ester
PubChem CID 74702
IUPAC Name ethyl 2-acetamidoacetate
SMILES CCOC(=O)CNC(=O)C

Specifications

Melting Point 47°C
Color White
Boiling Point 143°C
Quantity 1 g
Formula Weight 145.16
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetylglycine Ethyl Ester