missing translation for 'onlineSavingsMsg'
Learn More

N-Acetylglycine, 99%

Catalog No. AC102571000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC102571000 100 g Plastic bottle
AC102575000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC102571000 Supplier Thermo Scientific Chemicals Supplier No. 102571000

CAS: 543-24-8 | C4H7NO3 | 117.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 543-24-8
Molecular Formula C4H7NO3
Molecular Weight (g/mol) 117.1
MDL Number MFCD00004275
InChI Key OKJIRPAQVSHGFK-UHFFFAOYSA-N
Synonym n-acetylglycine, aceturic acid, acetamidoacetic acid, acetylglycine, ac-gly-oh, glycine, n-acetyl, acetylaminoacetic acid, acetylamino-acetic acid, acetylglycocoll, ethanoylaminoethanoic acid
PubChem CID 10972
ChEBI CHEBI:40410
IUPAC Name 2-acetamidoacetic acid
SMILES CC(=O)NCC(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 207.0°C to 209.0°C
Color White
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula CH3CONHCH2CO2H
Quantity 100 g
Beilstein 04, 354
Merck Index 15, 75
Solubility Information Solubility in water: 2.7% (15°C). Other solubilities: moderately soluble in alcohol, slightly soluble in acetone, chloroform and, glacial acetic acid, practically insoluble in ether and benzene
Formula Weight 117.1
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N-Acetylglycine, 99%
Show More Show Less
EINECSNumber 208-839-6
TSCA TSCA

RUO – Research Use Only