missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals N-Acetylglycine, 99%
CAS: 543-24-8 | C4H7NO3 | 117.104 g/mol
$63.55 - $180.05
Chemical Identifiers
| CAS | 543-24-8 |
|---|---|
| Molecular Formula | C4H7NO3 |
| Molecular Weight (g/mol) | 117.104 |
| MDL Number | MFCD00004275 |
| InChI Key | OKJIRPAQVSHGFK-UHFFFAOYSA-N |
| Synonym | n-acetylglycine, aceturic acid, acetamidoacetic acid, acetylglycine, ac-gly-oh, glycine, n-acetyl, acetylaminoacetic acid, acetylamino-acetic acid, acetylglycocoll, ethanoylaminoethanoic acid |
| PubChem CID | 10972 |
| ChEBI | CHEBI:40410 |
| IUPAC Name | 2-acetamidoacetic acid |
| SMILES | CC(=O)NCC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2188722
|
Thermo Scientific Chemicals
B2188722 |
100 g |
Each for $63.55
|
|
|||||
|
AAB2188736
|
Thermo Scientific Chemicals
B2188736 |
500 g |
Each for $180.05
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 543-24-8 | |
| 117.104 | |
| OKJIRPAQVSHGFK-UHFFFAOYSA-N | |
| 10972 | |
| 2-acetamidoacetic acid |
| C4H7NO3 | |
| MFCD00004275 | |
| n-acetylglycine, aceturic acid, acetamidoacetic acid, acetylglycine, ac-gly-oh, glycine, n-acetyl, acetylaminoacetic acid, acetylamino-acetic acid, acetylglycocoll, ethanoylaminoethanoic acid | |
| CHEBI:40410 | |
| CC(=O)NCC(=O)O |
Specifications
| ∼210°C (decomposition) | |
| C4H7NO3 | |
| 774114 | |
| n-acetylglycine, aceturic acid, acetamidoacetic acid, acetylglycine, ac-gly-oh, glycine, n-acetyl, acetylaminoacetic acid, acetylamino-acetic acid, acetylglycocoll, ethanoylaminoethanoic acid | |
| CC(=O)NCC(=O)O | |
| 100 g | |
| 10972 | |
| 117.1 | |
| N-Acetylglycine |
| 543-24-8 | |
| MFCD00004275 | |
| 14,80 | |
| OKJIRPAQVSHGFK-UHFFFAOYSA-N | |
| 2-acetamidoacetic acid | |
| 117.104 | |
| CHEBI:40410 | |
| 99% |
Safety and Handling
H500
EINECSNumber : 208-839-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only