missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetylglycine, 99%

Catalog No. AAB2188736
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2188736 500 g
AAB2188722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2188736 Supplier Thermo Scientific Chemicals Supplier No. B2188736

CAS: 543-24-8 | C4H7NO3 | 117.104 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 543-24-8
Molecular Formula C4H7NO3
Molecular Weight (g/mol) 117.104
MDL Number MFCD00004275
InChI Key OKJIRPAQVSHGFK-UHFFFAOYSA-N
Synonym n-acetylglycine, aceturic acid, acetamidoacetic acid, acetylglycine, ac-gly-oh, glycine, n-acetyl, acetylaminoacetic acid, acetylamino-acetic acid, acetylglycocoll, ethanoylaminoethanoic acid
PubChem CID 10972
ChEBI CHEBI:40410
IUPAC Name 2-acetamidoacetic acid
SMILES CC(=O)NCC(=O)O

Specifications

Melting Point ∼210°C (decomposition)
Beilstein 774114
Merck Index 14,80
Quantity 500 g
Formula Weight 117.1
Percent Purity 99%
Chemical Name or Material N-Acetylglycine
Hazard Category H500
EINECSNumber 208-839-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only