missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetylethylenediamine 95.0+%, TCI America™
$71.23 - $204.07
Chemical Identifiers
| CAS | 1001-53-2 |
|---|---|
| Molecular Formula | C4H10N2O |
| Molecular Weight (g/mol) | 102.137 |
| MDL Number | MFCD00008163 |
| InChI Key | DAKZISABEDGGSV-UHFFFAOYSA-N |
| Synonym | n-2-aminoethyl acetamide, n-acetylethylenediamine, acetamide, n-2-aminoethyl, n-2-amino-ethyl-acetamide, unii-fql33v3kzd, fql33v3kzd, acetylethylenediamine, 2-acetamidoethylamine, acetylethylene diamine, n-acetylethylendiamine |
| PubChem CID | 66082 |
| IUPAC Name | N-(2-aminoethyl)acetamide |
| SMILES | CC(=O)NCCN |
Chemical Identifiers
| 1001-53-2 | |
| 102.137 | |
| DAKZISABEDGGSV-UHFFFAOYSA-N | |
| 66082 | |
| CC(=O)NCCN |
| C4H10N2O | |
| MFCD00008163 | |
| n-2-aminoethyl acetamide, n-acetylethylenediamine, acetamide, n-2-aminoethyl, n-2-amino-ethyl-acetamide, unii-fql33v3kzd, fql33v3kzd, acetylethylenediamine, 2-acetamidoethylamine, acetylethylene diamine, n-acetylethylendiamine | |
| N-(2-aminoethyl)acetamide |
Specifications
| 1001-53-2 | |
| 143°C | |
| MFCD00008163 | |
| 3259 | |
| DAKZISABEDGGSV-UHFFFAOYSA-N | |
| N-(2-aminoethyl)acetamide | |
| 66082 | |
| ≥95.0% (GC) | |
| N-Acetylethylenediamine |
| White-Yellow | |
| C4H10N2O | |
| 5 g | |
| n-2-aminoethyl acetamide, n-acetylethylenediamine, acetamide, n-2-aminoethyl, n-2-amino-ethyl-acetamide, unii-fql33v3kzd, fql33v3kzd, acetylethylenediamine, 2-acetamidoethylamine, acetylethylene diamine, n-acetylethylendiamine | |
| CC(=O)NCCN | |
| 102.137 | |
| 102.14 | |
| Crystals |
Safety and Handling
EINECSNumber : (2)-0722
TSCA : No