missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-trans-4-hydroxy-L-proline, 98%, Thermo Scientific Chemicals
$86.88 - $86.88
Chemical Identifiers
| CAS | 33996-33-7 |
|---|---|
| Molecular Formula | C7H11NO4 |
| Molecular Weight (g/mol) | 173.168 |
| MDL Number | MFCD00037339 |
| InChI Key | BAPRUDZDYCKSOQ-RITPCOANSA-N |
| Synonym | oxaceprol, n-acetyl-l-hydroxyproline, 2s,4r-1-acetyl-4-hydroxypyrrolidine-2-carboxylic acid, oxaceprol inn, ac-hyp-oh, l-proline, 1-acetyl-4-hydroxy-, 4r, unii-q0xv76b96l, ac-hydroxyproline, trans-1-acetyl-4-hydroxy-l-proline, jonctum |
| PubChem CID | 65784 |
| IUPAC Name | (2S,4R)-1-acetyl-4-hydroxypyrrolidine-2-carboxylic acid |
| SMILES | CC(=O)N1CC(CC1C(=O)O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2376704
|
Thermo Scientific Chemicals
B2376704 |
2 g |
Each for $86.88
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 33996-33-7 | |
| 173.168 | |
| BAPRUDZDYCKSOQ-RITPCOANSA-N | |
| 65784 | |
| CC(=O)N1CC(CC1C(=O)O)O |
| C7H11NO4 | |
| MFCD00037339 | |
| oxaceprol, n-acetyl-l-hydroxyproline, 2s,4r-1-acetyl-4-hydroxypyrrolidine-2-carboxylic acid, oxaceprol inn, ac-hyp-oh, l-proline, 1-acetyl-4-hydroxy-, 4r, unii-q0xv76b96l, ac-hydroxyproline, trans-1-acetyl-4-hydroxy-l-proline, jonctum | |
| (2S,4R)-1-acetyl-4-hydroxypyrrolidine-2-carboxylic acid |
Specifications
| 131°C to 133°C | |
| White | |
| MFCD00037339 | |
| 14,6903 | |
| BAPRUDZDYCKSOQ-RITPCOANSA-N | |
| (2S,4R)-1-acetyl-4-hydroxypyrrolidine-2-carboxylic acid | |
| 173.168 | |
| 173.17 | |
| Crystalline Powder |
| 33996-33-7 | |
| C7H11NO4 | |
| 84043 | |
| oxaceprol, n-acetyl-l-hydroxyproline, 2s,4r-1-acetyl-4-hydroxypyrrolidine-2-carboxylic acid, oxaceprol inn, ac-hyp-oh, l-proline, 1-acetyl-4-hydroxy-, 4r, unii-q0xv76b96l, ac-hydroxyproline, trans-1-acetyl-4-hydroxy-l-proline, jonctum | |
| CC(=O)N1CC(CC1C(=O)O)O | |
| 2 g | |
| 65784 | |
| 99% | |
| N-Acetyl-trans-4-hydroxy-L-proline |
Safety and Handling
H318-H335-H315; Causes serious eye damage. May cause respiratory irritation. Causes skin irritation.
P280-P305+P351+P338-P310
H318
EINECSNumber : 251-780-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only