missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-trans-4-hydroxy-L-proline, 98%, Thermo Scientific Chemicals

Catalog No. p-7076520
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2376709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2376709 Supplier Thermo Scientific Chemicals Supplier No. B2376709
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33996-33-7
Molecular Formula C7H11NO4
Molecular Weight (g/mol) 173.168
MDL Number MFCD00037339
InChI Key BAPRUDZDYCKSOQ-RITPCOANSA-N
Synonym oxaceprol, n-acetyl-l-hydroxyproline, 2s,4r-1-acetyl-4-hydroxypyrrolidine-2-carboxylic acid, oxaceprol inn, ac-hyp-oh, l-proline, 1-acetyl-4-hydroxy-, 4r, unii-q0xv76b96l, ac-hydroxyproline, trans-1-acetyl-4-hydroxy-l-proline, jonctum
PubChem CID 65784
IUPAC Name (2S,4R)-1-acetyl-4-hydroxypyrrolidine-2-carboxylic acid
SMILES CC(=O)N1CC(CC1C(=O)O)O

Specifications

Melting Point 131°C to 133°C
Packaging Glass Bottle
Beilstein 84043
Quantity 10 g
Formula Weight 173.17
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-trans-4-hydroxy-L-proline
Hazard Category H318
Precautionary Statement P280-P305+P351+P338-P310
EINECSNumber 251-780-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only