missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-Leu-Glu-His-Asp-7-amino-4-methylcoumarin

$255.97 - $255.97

Chemical Identifiers

CAS 292633-16-0
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66335LB0
View Documents
Thermo Scientific Chemicals
J66335LB0
5mg
Each for $255.97
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

292633-16-0

Specifications

292633-16-0
Store at -20°C
711.73
5mg
White
Lyophilized Powder
N-Acetyl-Leu-Glu-His-Asp-7-amino-4-methylcoumarin
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only