missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-tyrosine Ethyl Ester Monohydrate 98.0+%, TCI America™

Catalog No. A01185G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A01185G 5 g
A01181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A01185G Supplier TCI America Supplier No. A01185G

Chemical Identifiers

CAS 36546-50-6
Molecular Formula C13H19NO5
Molecular Weight (g/mol) 269.30
MDL Number MFCD00149093
InChI Key YWAVLHZJMWEYTA-HVPILOLGNA-N
Synonym n-acetyl-l-tyrosine ethyl ester monohydrate, ethyl n-acetyl-l-tyrosinate hydrate, ac-tyr-oet.h2o, atee, ethyl 2s-2-acetamido-3-4-hydroxyphenyl propanoate hydrate, n-acetyl-l-tyrosineethylestermonohydrate, n-acetyltyrosine ethyl ester monohydrate, ksc491m4n, n-acetyl tyrosine ethyl ester mono hydrate, acetyl-l-tyrosine ethyl ester hydrate
PubChem CID 2723594
IUPAC Name ethyl (2S)-2-acetamido-3-(4-hydroxyphenyl)propanoate hydrate
SMILES O.CCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(C)=O

Specifications

Melting Point 81°C
Color White-Yellow
Quantity 5 g
Formula Weight 251.28
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-L-tyrosine Ethyl Ester Monohydrate