missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-L-tyrosine ethyl ester monohydrate, 97%
CAS: 36546-50-6 | C13H19NO5 | 269.30 g/mol
$99.90 - $360.12
Chemical Identifiers
| CAS | 36546-50-6 |
|---|---|
| Molecular Formula | C13H19NO5 |
| Molecular Weight (g/mol) | 269.30 |
| MDL Number | MFCD00149093 |
| InChI Key | YWAVLHZJMWEYTA-HVPILOLGNA-N |
| Synonym | n-acetyl-l-tyrosine ethyl ester monohydrate, ethyl n-acetyl-l-tyrosinate hydrate, ac-tyr-oet.h2o, atee, ethyl 2s-2-acetamido-3-4-hydroxyphenyl propanoate hydrate, n-acetyl-l-tyrosineethylestermonohydrate, n-acetyltyrosine ethyl ester monohydrate, ksc491m4n, n-acetyl tyrosine ethyl ester mono hydrate, acetyl-l-tyrosine ethyl ester hydrate |
| PubChem CID | 2723594 |
| IUPAC Name | ethyl (2S)-2-acetamido-3-(4-hydroxyphenyl)propanoate;hydrate |
| SMILES | O.CCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(C)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1157706
|
Thermo Scientific Chemicals
A1157706 |
5 g |
Each for $99.90
|
|
|||||
|
AAA1157714
|
Thermo Scientific Chemicals
A1157714 |
25 g |
Each for $360.12
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 36546-50-6 | |
| 269.30 | |
| YWAVLHZJMWEYTA-HVPILOLGNA-N | |
| 2723594 | |
| O.CCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(C)=O |
| C13H19NO5 | |
| MFCD00149093 | |
| n-acetyl-l-tyrosine ethyl ester monohydrate, ethyl n-acetyl-l-tyrosinate hydrate, ac-tyr-oet.h2o, atee, ethyl 2s-2-acetamido-3-4-hydroxyphenyl propanoate hydrate, n-acetyl-l-tyrosineethylestermonohydrate, n-acetyltyrosine ethyl ester monohydrate, ksc491m4n, n-acetyl tyrosine ethyl ester mono hydrate, acetyl-l-tyrosine ethyl ester hydrate | |
| ethyl (2S)-2-acetamido-3-(4-hydroxyphenyl)propanoate;hydrate |
Specifications
| 80°C to 81°C | |
| C13H19NO5 | |
| n-acetyl-l-tyrosine ethyl ester monohydrate, ethyl n-acetyl-l-tyrosinate hydrate, ac-tyr-oet.h2o, atee, ethyl 2s-2-acetamido-3-4-hydroxyphenyl propanoate hydrate, n-acetyl-l-tyrosineethylestermonohydrate, n-acetyltyrosine ethyl ester monohydrate, ksc491m4n, n-acetyl tyrosine ethyl ester mono hydrate, acetyl-l-tyrosine ethyl ester hydrate | |
| O.CCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(C)=O | |
| 5 g | |
| 2723594 | |
| Moisture sensitive | |
| N-Acetyl-L-tyrosine ethyl ester monohydrate |
| 36546-50-6 | |
| MFCD00149093 | |
| YWAVLHZJMWEYTA-HVPILOLGNA-N | |
| ethyl (2S)-2-acetamido-3-(4-hydroxyphenyl)propanoate;hydrate | |
| 269.30 | |
| 269.30 (251.28 Anhydrous) | |
| 97% |
Safety and Handling
EINECSNumber : 212-663-5
TSCA : Yes
Recommended Storage : Store at -20°C; Store under Nitrogen
RUO – Research Use Only