missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-tryptophan methyl ester, 95%

CAS: 2824-57-9 | C14H16N2O3 | 260.29 g/mol

Supplier:  Thermo Scientific Chemicals H6693714

 View more versions of this product

Catalog No. AAH6693714

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Acetyl-L-tryptophan methyl ester
White
MFCD00022759
XZECNVJPYDPBAM-ZDUSSCGKSA-N
methyl (2S)-2-acetamido-3-(1H-indol-3-yl)propanoate
260.29
260.29
Powder
2824-57-9
C14H16N2O3
methyl n-acetyl-l-tryptophanate, ac-trp-ome, methyl acetyltryptophanate, l-tryptophan, n-acetyl-, methyl ester, unii-mup4y18zq5, n-acetyltryptophan methyl ester, n-acetyl-l-tryptophan methyl ester, mup4y18zq5, methyl 2s-2-acetamido-3-1h-indol-3-yl propanoate, l-tryptophan,n-acetyl-, methyl ester
COC(=O)[C@H](CC1=CNC2=CC=CC=C12)NC(C)=O
25 g
102337
95%

Chemical Identifiers

2824-57-9
260.29
XZECNVJPYDPBAM-ZDUSSCGKSA-N
102337
COC(=O)[C@H](CC1=CNC2=CC=CC=C12)NC(C)=O
C14H16N2O3
MFCD00022759
methyl n-acetyl-l-tryptophanate, ac-trp-ome, methyl acetyltryptophanate, l-tryptophan, n-acetyl-, methyl ester, unii-mup4y18zq5, n-acetyltryptophan methyl ester, n-acetyl-l-tryptophan methyl ester, mup4y18zq5, methyl 2s-2-acetamido-3-1h-indol-3-yl propanoate, l-tryptophan,n-acetyl-, methyl ester
methyl (2S)-2-acetamido-3-(1H-indol-3-yl)propanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only