missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-tryptophan, 99%

Catalog No. p-7042876
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6409606 5 g
AAJ6409614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6409606 Supplier Thermo Scientific Chemicals Supplier No. J6409606
Only null left

CAS: 1218-34-4 | C13H14N2O3 | 246.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1218-34-4
Molecular Formula C13H14N2O3
Molecular Weight (g/mol) 246.27
MDL Number MFCD00065976
InChI Key DZTHIGRZJZPRDV-LBPRGKRZSA-N
Synonym n-acetyl-l-tryptophan, ac-trp-oh, acetyl-l-tryptophan, acetyltryptophan, acetyl-l-trp, ac-try, s-n-acetyltryptophan, unii-u9264t8oae, tryptophan, n-acetyl, s-2-acetamido-3-1h-indol-3-yl propanoic acid
PubChem CID 700653
ChEBI CHEBI:74640
IUPAC Name (2S)-2-acetamido-3-(1H-indol-3-yl)propanoic acid
SMILES CC(=O)N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

Melting Point 186°C
Color White
Quantity 5 g
Formula Weight 246.3
Optical Rotation +26° (c=1 in 1N NaOH)
Percent Purity 99%
Physical Form Powder
Chemical Name or Material N-Acetyl-L-tryptophan
EINECSNumber 214-935-9
RTECSNumber YN6160000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only