missing translation for 'onlineSavingsMsg'
Learn More

N-acetyl-l-proline, 99%

Catalog No. p-3212933
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC376640010 1 g Glass bottle
AC376640050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC376640010 Supplier Thermo Scientific Chemicals Supplier No. 376640010

CAS: 68-95-1 | C7H11NO3 | 157.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68-95-1
Molecular Formula C7H11NO3
Molecular Weight (g/mol) 157.17
MDL Number MFCD00020837
InChI Key GNMSLDIYJOSUSW-UHFFFAOYNA-N
Synonym n-acetyl-l-proline, acetylproline, ac-pro-oh, n-acetylproline, 1-acetyl-l-proline, acetyl proline, l-proline, 1-acetyl, acetyl-l-proline, 2s-1-acetylpyrrolidine-2-carboxylic acid, s-1-acetylpyrrolidine-2-carboxylic acid
PubChem CID 66141
ChEBI CHEBI:21560
IUPAC Name (2S)-1-acetylpyrrolidine-2-carboxylic acid
SMILES CC(=O)N1CCCC1C(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 115°C to 117°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Specific Rotation Condition −86° (20°C c=1 EtOH)
Specific Rotation -86
Formula Weight 157.17
Percent Purity 99%
Physical Form Powder
Chemical Name or Material N-Acetyl-L-proline
Show More Show Less
EINECSNumber 200-698-9

RUO – Research Use Only