missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-L-proline, 99%

Catalog No. aal1430006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1430006 5 g
AAL1430003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1430006 Supplier Thermo Scientific Chemicals Supplier No. L1430006

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68-95-1
Molecular Formula C7H11NO3
Molecular Weight (g/mol) 157.17
MDL Number MFCD00020837
InChI Key GNMSLDIYJOSUSW-UHFFFAOYNA-N
Synonym n-acetyl-l-proline, acetylproline, ac-pro-oh, n-acetylproline, 1-acetyl-l-proline, acetyl proline, l-proline, 1-acetyl, acetyl-l-proline, 2s-1-acetylpyrrolidine-2-carboxylic acid, s-1-acetylpyrrolidine-2-carboxylic acid
PubChem CID 66141
ChEBI CHEBI:21560
IUPAC Name (2S)-1-acetylpyrrolidine-2-carboxylic acid
SMILES CC(=O)N1CCCC1C(O)=O

Specifications

Melting Point 117°C to 118°C
Beilstein 83200
Quantity 5 g
Formula Weight 157.18
Optical Rotation −116° (c=1 in Water)
Percent Purity 99%
Chemical Name or Material N-Acetyl-L-proline
EINECSNumber 200-698-9
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only