missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-phenylalanine 99.0+%, TCI America™

Catalog No. p-7117504
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15415G 5 g
A154125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15415G Supplier TCI America Supplier No. A15415G
Only null left

Chemical Identifiers

CAS 2018-61-3
Molecular Formula C11H13NO3
Molecular Weight (g/mol) 207.23
MDL Number MFCD00063158
InChI Key CBQJSKKFNMDLON-JTQLQIEISA-N
Synonym n-acetyl-l-phenylalanine, ac-phe-oh, acetyl-l-phenylalanine, acetylphenylalanine, l-n-acetylphenylalanine, s-2-acetamido-3-phenylpropanoic acid, phenylalanine, n-acetyl, n-acetyl-3-phenyl-l-alanine, unii-np5bt39467, 2s-2-acetamido-3-phenylpropanoic acid
PubChem CID 74839
ChEBI CHEBI:16259
IUPAC Name (2S)-2-acetamido-3-phenylpropanoic acid
SMILES CC(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 168°C
Color White
Quantity 5 g
Formula Weight 207.23
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-L-phenylalanine
EINECSNumber (9)-1617
TSCA Yes