missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-methionine 98.0+%, TCI America™

Supplier:  TCI America A20565G

 View more versions of this product

Catalog No. A20565G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

N-Acetyl-L-methionine
105°C
C7H13NO3S
5 g
XUYPXLNMDZIRQH-LURJTMIESA-N
(2S)-2-acetamido-4-(methylsulfanyl)butanoic acid
448580
191.25
Crystalline Powder
65-82-7
White-Yellow
MFCD00064441
n-acetyl-l-methionine, n-acetylmethionine, acetyl-l-methionine, ac-met-oh, l-n-acetyl methionine, acetylmethionine, methionamine, methionine, n-acetyl-, l, thiomedon, l-methionine, n-acetyl
CSCC[C@H](NC(C)=O)C(O)=O
191.25
CHEBI:21557
≥98.0% (HPLC,T)

Chemical Identifiers

65-82-7
191.25
XUYPXLNMDZIRQH-LURJTMIESA-N
448580
(2S)-2-acetamido-4-(methylsulfanyl)butanoic acid
C7H13NO3S
MFCD00064441
n-acetyl-l-methionine, n-acetylmethionine, acetyl-l-methionine, ac-met-oh, l-n-acetyl methionine, acetylmethionine, methionamine, methionine, n-acetyl-, l, thiomedon, l-methionine, n-acetyl
CHEBI:21557
CSCC[C@H](NC(C)=O)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : PD0480000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions