missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-L-histidine monohydrate

Catalog No. AAJ6565703
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6565703 1 g
AAJ6565706 5 g
AAJ6565714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6565703 Supplier Thermo Scientific Chemicals Supplier No. J6565703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 39145-52-3 | C8H13N3O4 | 215.209 g/mol

N-acetylhistidine, a novel osmolyte in the lens of Atlantic salmon. An ectotherm homologue of human predicted gene NAT16 encodes histidine N-acetyltransferase responsible for NÃŽ±-acetylhistidine synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-acetylhistidine, a novel osmolyte in the lens of Atlantic salmon. An ectotherm homologue of human predicted gene NAT16 encodes histidine N-acetyltransferase responsible for Nα-acetylhistidine synthesis.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 39145-52-3
Molecular Formula C8H13N3O4
Molecular Weight (g/mol) 215.209
MDL Number MFCD00149320
InChI Key PSWSDQRXCOJSFC-FJXQXJEOSA-N
Synonym ac-his-oh.h2o, n-acetyl-l-histidine hydrate, n-acetyl-l-histidine monohydrate, unii-pq2tc3x11o, s-2-acetamido-3-1h-imidazol-4-yl propanoic acid hydrate, pq2tc3x11o, acetyl histidine, 2s-2-acetylamino-3-1h-imidazol-4-yl propanoic acid hydrate, l-n-acetylhistidine monohydrate
PubChem CID 2724380
IUPAC Name (2S)-2-acetamido-3-(1H-imidazol-5-yl)propanoic acid;hydrate
SMILES CC(=O)NC(CC1=CN=CN1)C(=O)O.O

Specifications

Melting Point 163°C
Color White
Solubility Information Soluble in water.
Quantity 1 g
Formula Weight 215.21 (197.09 Anhydrous)
Optical Rotation +47° (c=1 in Water)
Physical Form Powder
Chemical Name or Material N-Acetyl-L-histidine monohydrate
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only