missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-glutamine, 97%, Thermo Scientific™

Catalog No. p-4813568
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC451640050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC451640050 Supplier Thermo Scientific Chemicals Supplier No. 451640050
Only null left

Chemical Identifiers

CAS 35305-74-9
Molecular Formula C7H12N2O4
Molecular Weight (g/mol) 188.18
MDL Number MFCD00038159
InChI Key KSMRODHGGIIXDV-YFKPBYRVSA-N
Synonym n-acetyl-l-glutamine, aceglutamide, l-glutamine, n-acetyl, nalpha-acetyl-l-glutamine, s-2-acetamido-5-amino-5-oxopentanoic acid, acetylglutamine, n-alpha-acetyl-l-glutamine, n-acetylglutamine, acutil-s, acetyl-l-glutamine
PubChem CID 182230
ChEBI CHEBI:21553
SMILES CC(=O)N[C@@H](CCC(N)=O)C(O)=O

Specifications

Melting Point 206°C
Ignition Residue 0.2% max
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: soluble
IUPAC Name (2S)-4-carbamoyl-2-acetamidobutanoic acid
Formula Weight 188.18
Percent Purity 97%
Chemical Name or Material N-Acetyl-L-glutamine

RUO – Research Use Only