missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-glutamic Acid 98.0+%, TCI America™

Supplier:  TCI America A069325G

 View more versions of this product

Catalog No. A069325G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

N-Acetyl-L-glutamic Acid
198°C
C7H11NO5
25 g
RFMMMVDNIPUKGG-YFKPBYRVSA-N
(2S)-2-acetamidopentanedioic acid
70914
189.17
Crystalline Powder
1188-37-0
White
MFCD00002802
n-acetyl-l-glutamic acid, acetylglutamic acid, n-acetyl-l-glutamate, n-acetylglutamic acid, n-acetylglutamate, acetyl glutamic acid, acetyl-l-glutamic acid, l-glutamic acid, n-acetyl, l-n-acetylglutamic acid, ac-glu-oh
CC(=O)NC(CCC(=O)O)C(=O)O
189.167
CHEBI:17533
≥98.0% (HPLC,T)

Chemical Identifiers

1188-37-0
189.167
RFMMMVDNIPUKGG-YFKPBYRVSA-N
70914
(2S)-2-acetamidopentanedioic acid
C7H11NO5
MFCD00002802
n-acetyl-l-glutamic acid, acetylglutamic acid, n-acetyl-l-glutamate, n-acetylglutamic acid, n-acetylglutamate, acetyl glutamic acid, acetyl-l-glutamic acid, l-glutamic acid, n-acetyl, l-n-acetylglutamic acid, ac-glu-oh
CHEBI:17533
CC(=O)NC(CCC(=O)O)C(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-2029

RTECSNumber : LZ9725000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions